missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Galectin-3-binding Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Galectin-3-binding protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6296-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022260

  • 770.00€ / 100µg
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_620796.1
Galectin-3-binding Protein
Research Use Only
Lgals3bp
Recombinant Protein
E. coli
AKDGDMRLVN GASASEGRVE IFYRGRWGTV CDNLWNLLDA HVVCRALGYE NATQALSRAA FGPGKGPIML DEVECTGNES SLANCSSLGW MVSHCGHEKD AGVVCSNDSR GIHILDLSGE LPDALGQIFD SQQDCDLFIQ VTGQGHGDLS LCAHTLILRT NPEAQALWQV VGSSVIMRVD AECMPVVRDF LRYFYSRRIE VSMSSVKCLH KLASAYGATE LQGYCGRLFV TLLPQDPTFH TPLELYEYAQ ATGDSVLEDL CVQFLAWNFE PLTQAEAWLS VPNALIQALL PKSELAVSSE LDLLKAVDQW STATGASHGD VERLVEQIRF PMMLPQELFE LQFNLSLYQG HQALFQRKTM EALEFHTVPL KVLAKYRSLN LTEDVYKPRL YTSSTWSSLL MAGAWSTQSY KYRQFYTYNY GSQSRYSSYQ NFQTPQHPSF LFKDKLISWS ATYLPTIQSC WNYGFSCTSD ELPVLGLTTS SYSDPTIGYE NKALILCGGY SVVDVTTFIG SKAPIPGTQE TNSSKTPSLF PCASGAFSSF RVVIRPFYLT NSTDTE
Greater than 90% as determined by SDS-PAGE.
245955
100 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers