missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Galectin-3-binding Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Galectin-3-binding protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6296-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_620796.1 | |
Galectin-3-binding Protein | |
Research Use Only | |
Lgals3bp | |
Recombinant Protein | |
E. coli | |
AKDGDMRLVN GASASEGRVE IFYRGRWGTV CDNLWNLLDA HVVCRALGYE NATQALSRAA FGPGKGPIML DEVECTGNES SLANCSSLGW MVSHCGHEKD AGVVCSNDSR GIHILDLSGE LPDALGQIFD SQQDCDLFIQ VTGQGHGDLS LCAHTLILRT NPEAQALWQV VGSSVIMRVD AECMPVVRDF LRYFYSRRIE VSMSSVKCLH KLASAYGATE LQGYCGRLFV TLLPQDPTFH TPLELYEYAQ ATGDSVLEDL CVQFLAWNFE PLTQAEAWLS VPNALIQALL PKSELAVSSE LDLLKAVDQW STATGASHGD VERLVEQIRF PMMLPQELFE LQFNLSLYQG HQALFQRKTM EALEFHTVPL KVLAKYRSLN LTEDVYKPRL YTSSTWSSLL MAGAWSTQSY KYRQFYTYNY GSQSRYSSYQ NFQTPQHPSF LFKDKLISWS ATYLPTIQSC WNYGFSCTSD ELPVLGLTTS SYSDPTIGYE NKALILCGGY SVVDVTTFIG SKAPIPGTQE TNSSKTPSLF PCASGAFSSF RVVIRPFYLT NSTDTE | |
Greater than 90% as determined by SDS-PAGE. |
245955 | |
200 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |