missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant RABV NucleoProtein
A DNA sequence encoding the Rabies virus (strain PM) (RABV) Nucleoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7060-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
RABV NucleoProtein | |
Research Use Only | |
N | |
Recombinant Protein | |
Yeast | |
MDADKIVFKV NNQVVSLKPE IIVDQYEYKY PAIKDLKKPC ITLGKAPDLN KAYKSVLSGM NAAKLDPDDV CSYLAAAMQF FEGTCPEDWT SYGILIARKG DRITPNSLVE IKRTDVEGNW ALTGGMELTR DPTVSEHASL VGLLLSLYRL SKISGQNTGN YKTNIADRIE QIFETAPFVK IVEHHTLMTT HKMCANWSTI PNFRFLAGTY DMFFSRIEHL YSAIRVGTVV TAYEDCSGLV SFTGFIKQIN LTAREAILYF FHKNFEEEIR RMFEPGQETA VPHSYFIHFR SLGLSGKSPY SSNAVGHVFN LIHFVGCYMG QVRSLNATVI AACAPHEMSV LGGYLGEEFF GKGTFERRFF RDEKELQEYE AAELTKTDVA LADDGTVNSD DEDYFSGETR SPEAVYTRIM MNGGRLKRSH IRRYVSVSSN HQARPNSFAE FLNKTYSNDS | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
RABV | |
His | |
Tris-based buffer, 50% glycerol |