missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant RABV GlycoProtein

A DNA sequence encoding the Rabies virus (strain PM) (RABV) Glycoprotein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7425-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028810

  • 985.00€ / 100µg
Estimated Shipment:
to see stock.
Add to basket

Description

Description

Specifications

Specifications

RABV GlycoProtein
Research Use Only
G
Recombinant Protein
E. coli
KFPIYTIPDK LGPWSPIDIH HLSCPNNLVV EDEGCTNLSG FSYMELKVGY ISAIKVNGFT CTGVVTEAET YTNFVGYVTT TFKRKHFRPT PDACRAAYNW KMAGDPRYEE SLHNPYPDYH WLRTVKTTKE SLVIISPSVA DLDPYDKSLH SRVFPSGKCS GITISSTYCS TNHDYTIWMP ENPRLGTSCD IFTNSRGKRA SKGGKTCGFV DERGLYKSLK GACKLKLCGV LGLRLMDGTW VAMQTSDETK WCPPDQLVNL HDFRSDEIEH LVVEELVKKR EECLDALESI MATKSVSFRR LSHLRKLVPG FGKAYTIFNK TLMEADAHYK SVRTWNEIIP SKGCLRVGGR CHPHVNGVFF NGIILGPDGH VLIPEMQSSL LQQHMELLES SVIPLMHPLA DPSTVFKDGD EAEDFVEVHL PDVHKQISGV DLGLPSWGKY
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
RABV
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers