missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig E-Selectin / CD62e / SELE Protein
A DNA sequence encoding the Sus scrofa (Pig) E-Selectin / CD62e / SELE, was expressed in the hosts and tags indicated.
352.00€ - 3175.00€
Specifications
Accession Number | NP_999433.1 |
---|---|
Gene ID (Entrez) | 397508 |
Name | E-Selectin / CD62e / SELE Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16084890
|
enQuireBio™
QP6661-YE-100UG |
100 μg |
1100.00€
100µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16024900
|
enQuireBio™
QP6661-YE-500UG |
500 μg |
2650.00€
500µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16094890
|
enQuireBio™
QP6661-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16014900
|
enQuireBio™
QP6661-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16034900
|
enQuireBio™
QP6661-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16004900
|
enQuireBio™
QP6661-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_999433.1 | |
E-Selectin / CD62e / SELE Protein | |
Not determined. | |
Extracellular Domain | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |
397508 | |
Research Use Only | |
SELE | |
Recombinant Protein | |
Yeast | |
WSYSASTETM TFDDASAYCQ QRYTHLVAIQ NHAEIEYLNS TFNYSASYYW IGIRKINGTW TWIGTKKALT PEATNWAPGE PNNKQSNEDC VEIYIKRDKD SGKWNDERCS KKKLALCYTA ACTPTSCSGH GECIETINSS TCQCYPGFRG LQCEQVVECD ALENPVNGVV TCPQSLPWNT TCAFECKEGF ELIGPEHLQC TSSGSWDGKK PTCKAVTCDT VGHPQNGDVS CNHSSIGEFA YKSTCHFTCA EGFGLQGPAQ IECTAQGQWT QQAPVCKAVK CPAVSQPKNG LVKFTHSPTG EFTYKSSCAF SCEEGFELRG SAQLACTSQG QWTQEVPSCQ VVQCSSLEVP REINMSCSGE PVFGAVCTFA CPEGWMLNGS VALTCGATGH WSGMLPTCEA PAESKIP | |
Greater than 90% as determined by SDS-PAGE. |