missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant P. aeruginosa Beta-lactamase Protein
A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) Beta-lactamase, was expressed in the hosts and tags indicated. option, ab
Brand: enQuireBio™ QP7119-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_252799.1 | |
P. aeruginosa Beta-lactamase Protein | |
Research Use Only | |
ampC | |
Recombinant Protein | |
E. coli | |
GEAPADRLKA LVDAAVQPVM KANDIPGLAV AISLKGEPHY FSYGLASKED GRRVTPETLF EIGSVSKTFT ATLAGYALTQ DKMRLDDRAS QHWPALQGSR FDGISLLDLA TYTAGGLPLQ FPDSVQKDQA QIRDYYRQWQ PTYAPGSQRL YSNPSIGLFG YLAARSLGQP FERLMEQQVF PALGLEQTHL DVPEAALAQY AQGYGKDDRP LRVGPGPLDA EGYGVKTSAA DLLRFVDANL HPERLDRPWA QALDATHRGY YKVGDMTQGL GWEAYDWPIS LKRLQAGNST PMALQPHRIA RLPAPQALEG QRLLNKTGST NGFGAYVAFV PGRDLGLVIL ANRNYPNAER VKIAYAILSG LEQQGKVPLK R | |
Greater than 90% as determined by SDS-PAGE. |
878149 | |
100 μg | |
Not determined. | |
Full Length | |
P. aeruginosa | |
His-SUMO | |
Tris-based buffer, 50% glycerol |