missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse WNT1-inducible-signaling pathway protein 1

A DNA sequence encoding the Mus musculus (Mouse) WNT1-inducible-signaling pathway protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8274-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16043701

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_058569.2
WNT1-inducible-signaling pathway protein 1
Research Use Only
WISP2
Recombinant Protein
E. coli
QLCPAPCACP WTPPQCPPGV PLVLDGCGCC RVCARRLGES CDHLHVCDPS QGLVCQPGAG PSGRGAVCLF EEDDGSCEVN GRRYLDGETF KPNCRVLCRC DDGGFTCLPL CSEDVRLPSW DCPRPRRIQV PGRCCPEWVC DQAVMQPAIQ PSSAQGHQLS ALVTPASADG PCPNWSTAWG PCSTTCGLGI ATRVSNQNRF CQLEIQRRLC LSRPCLASRS HGSWNSAF
Greater than 90% as determined by SDS-PAGE.
22403
200 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers