missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Uromodulin Protein
A DNA sequence encoding the Mus musculus (Mouse) Uromodulin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9459-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001265534.1 | |
Uromodulin Protein | |
Research Use Only | |
Umod | |
Recombinant Protein | |
Yeast | |
NSTEARRCSE CHNNATCTVD GVVTTCSCQT GFTGDGLVCE DMDECATPWT HNCSNSSCVN TPGSFKCSCQ DGFRLTPELS CTDVDECSEQ GLSNCHALAT CVNTEGDYLC VCPEGFTGDG WYCECSPGSC EPGLDCLPQG PDGKLVCQDP CNTYETLTEY WRSTEYGVGY SCDAGLHGWY RFTGQGGVRM AETCVPVLRC NTAAPMWLNG SHPSSSEGIV SRTACAHWSD QCCRWSTEIQ VKACPGGFYI YNLTAPPECN LAYCTDPSSV EGTCEECRVD EDCISDNGRW RCQCKQDSNI TDVSQLEYRL ECGANDIKMS LRKCQLQSLG FMNVFMYLND RQCSGFSESD ERDWMSIVTP ARNGPCGTVL RRNETHATYS NTLYLANAII IRDIIIRMNF ECSYPLDMKV SLKTSLQPMV SALNISLGGT GKFTVRMALF QSPTYTQPHQ GPSVMLSTEA FLYVGTMLDG GDLSRFVLLM TNCYATPSSN STDPVKYFII QDSCPRTEDT TIQVTENGES SQARFSVQMF RFAGNYDLVY LHCEVYLCDS TSEQCKPTCS GTRF | |
Greater than 90% as determined by SDS-PAGE. |
22242 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |