missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Troponin I, cardiac muscle Protein
A DNA sequence encoding the Mus musculus (Mouse) Troponin I, cardiac muscle, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7213-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_033432.1 | |
Troponin I, cardiac muscle Protein | |
Research Use Only | |
Tnni3 | |
Recombinant Protein | |
E. coli | |
ADESSDAAGE PQPAPAPVRR RSSANYRAYA TEPHAKKKSK ISASRKLQLK TLMLQIAKQE MEREAEERRG EKGRVLRTRC QPLELDGLGF EELQDLCRQL HARVDKVDEE RYDVEAKVTK NITEIADLTQ KIYDLRGKFK RPTLRRVRIS ADAMMQALLG TRAKESLDLR AHLKQVKKED IEKENREVGD WRKNIDALSG MEGRKKKFEG | |
Greater than 82.9% as determined by SDS-PAGE. |
21954 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |