missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse TIMP-1 / TIMP1 Protein

A DNA sequence encoding the Mus musculus (Mouse) TIMP-1 / TIMP1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6795-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16095900

  • 770.00€ / 100µg
Estimated Shipment: 02-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001037849.1
TIMP-1 / TIMP1 Protein
Research Use Only
Timp1
Recombinant Protein
E. coli
CSCAPPHPQT AFCNSDLVIR AKFMGSPEIN ETTLYQRYKI KMTKMLKGFK AVGNAADIRY AYTPVMESLC GYAHKSQNRS EEFLITGRLR NGNLHISACS FLVPWRTLSP AQQRAFSKTY SAGCGVCTVF PCLSIPCKLE SDTHCLWTDQ VLVGSEDYQS RHFACLPRNP GLCTWRSLGA R
Greater than 90% as determined by SDS-PAGE.
21857
100 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers