missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Spermine oxidase Protein
A DNA sequence encoding the Mus musculus (Mouse) Spermine oxidase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6701-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001171304.1 | |
Spermine oxidase Protein | |
Research Use Only | |
Smox | |
Recombinant Protein | |
Yeast | |
MQSCESSGDS ADDPLSRGLR RRGQPRVVVI GAGLAGLAAA RALLEQGFTD VTVLEASSHI GGRVQSVRLG DTTFELGATW IHGSHGNPIY QLAEANGLLE ETTDGERSVG RISLYSKNGV ACYLTNRGCR IPKDVVEEFS DLYNEVYNMT QEFFRHGKPV NAESQNSVGV FTREKVRNRI RDDPDDTEAT KRLKLAMIQQ YLKVESCESS SHSIDEVSLS AFGEWTEIPG AHHIIPSGFM RVVELLAEGI PPHVIQLGKP VRCIHWDQAS AHPRGPEIEP RGEGDHNHDT GEGGQSGENP QQGRWDEDEP WPVVVECEDC EVIPADHVIV TVSLGVLKRQ YTSFFRPCLP TEKVAAIHRL GIGTTDKIFL EFEEPFWGPE CNSLQFVWED EAESCTLTYP PELWYRKICG FDVLYPPERY GHVLSGWICG EEALVMERCD DEAVAEICTE MLRQFTGNPN IPKPRRILRS AWGSNPYFRG SYSYTQVGSS GADVEKLAKP LPYTESSKTA PMQVLFSGEA THRKYYSTTH GALLSGQREA ARLIEMYRDL FQQGP | |
Greater than 90% as determined by SDS-PAGE. |
228608 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |