missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse SIGIRR / TIR8 Protein

A DNA sequence encoding the Mus musculus (Mouse) SIGIRR / TIR8, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8219-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16043361

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_075546.2
SIGIRR / TIR8 Protein
Research Use Only
Sigirr
Recombinant Protein
Yeast
MAGVCDMAPN FLSPSEDQAL GLALGREVAL NCTAWVFSRP QCPQPSVQWL KDGLALGNGS HFSLHEDFWV SANFSEIVSS VLVLNLTNAE DYGTFTCSVW NVSSHSFTLW RAGPAGH
Greater than 90% as determined by SDS-PAGE.
24058
1 mg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers