missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse RANKL / OPGL / TNFSF11 / CD254 Protein
A DNA sequence encoding the Mus musculus (Mouse) RANKL / OPGL / TNFSF11 / CD254, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6818-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_035743.2 | |
RANKL / OPGL / TNFSF11 / CD254 Protein | |
Research Use Only | |
Tnfsf11 | |
Recombinant Protein | |
E. coli | |
YFRAQMDPNR ISEDSTHCFY RILRLHENAD LQDSTLESED TLPDSCRRMK QAFQGAVQKE LQHIVGPQRF SGAPAMMEGS WLDVAQRGKP EAQPFAHLTI NAASIPSGSH KVTLSSWYHD RGWAKISNMT LSNGKLRVNQ DGFYYLYANI CFRHHETSGS VPTDYLQLMV YVVKTSIKIP SSHNLMKGGS TKNWSGNSEF HFYSINVGGF FKLRAGEEIS IQVSNPSLLD PDQDATYFGA FKVQDID | |
Greater than 90% as determined by SDS-PAGE. |
21943 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |