missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PLBD2 Protein
A DNA sequence encoding the Mus musculus (Mouse) PLBD2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7649-ma-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_076114.2 | |
PLBD2 Protein | |
Research Use Only | |
Plbd2 | |
Recombinant Protein | |
Mammal | |
LPTLGPGWQR QNPDPPVSRT RSLLLDAASG QLRLEDGFHP DAVAWANLTN AIRETGWAYL DLSTNGRYND SLQAYAAGVV EASVSEELIY MHWMNTVVNY CGPFEYEVGY CEKLKNFLEA NLEWMQREME LNPDSPYWHQ VRLTLLQLKG LEDSYEGRLT FPTGRFTIKP LGFLLLQISG DLEDLEPALN KTNTKPSLGS GSCSALIKLL PGGHDLLVAH NTWNSYQNML RIIKKYRLQF REGPQEEYPL VAGNNLVFSS YPGTIFSGDD FYILGSGLVT LETTIGNKNP ALWKYVQPQG CVLEWIRNVV ANRLALDGAT WADVFKRFNS GTYNNQWMIV DYKAFLPNGP SPGSRVLTIL EQIPGMVVVA DKTAELYKTT YWASYNIPYF ETVFNASGLQ ALVAQYGDWF SYTKNPRAKI FQRDQSLVED MDAMVRLMRY NDFLHDPLSL CEACNPKPNA ENAISARSDL NPANGSYPFQ ALHQRAHGGI DVKVTSFTLA KYMSMLAASG PTWDQCPPFQ WSKSPFHSML HMGQPDLWMF SPIRVPWD | |
Greater than 90% as determined by SDS-PAGE. |
71772 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |