missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Nuclease-sensitive element-binding protein 1

A DNA sequence encoding the Mus musculus (Mouse) Nuclease-sensitive element-binding protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9471-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16098381

  • 2195.00€ / 500µg
Estimated Shipment: 26-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_035862.2
Nuclease-sensitive element-binding protein 1
Research Use Only
Ybx1
Recombinant Protein
Yeast
SSEAETQQPP AAPAAALSAA DTKPGSTGSG AGSGGPGGLT SAAPAGGDKK VIATKVLGTV KWFNVRNGYG FINRNDTKED VFVHQTAIKK NNPRKYLRSV GDGETVEFDV VEGEKGAEAA NVTGPGGVPV QGSKYAADRN HYRRYPRRRG PPRNYQQNYQ NSESGEKNEG SESAPEGQAQ QRRPYRRRRF PPYYMRRPYA RRPQYSNPPV QGEVMEGADN QGAGEQGRPV RQNMYRGYRP RFRRGPPRQR QPREDGNEED KENQGDETQG QQPPQRRYRR NFNYRRRRPE NPKPQDGKET KAADPPAENS SAPEAEQGGA E
Greater than 90% as determined by SDS-PAGE.
22608
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers