missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse NKX2.2 Protein

A DNA sequence encoding the Mus musculus (Mouse) NKX2.2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6423-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16063210

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_035049.1
NKX2.2 Protein
Research Use Only
Nkx2-2
Recombinant Protein
E. coli
MSLTNTKTGF SVKDILDLPD TNDEDGSVAE GPEEESEGPE PAKRAGPLGQ GALDAVQSLP LKSPFYDSSD NPYTRWLAST EGLQYSLHGL AASAPPQDSS SKSPEPSADE SPDNDKETQG GGGDAGKKRK RRVLFSKAQT YELERRFRQQ RYLSAPEREH LASLIRLTPT QVKIWFQNHR YKMKRARAEK GMEVTPLPSP RRVAVPVLVR DGKPCHALKA QDLAAATFQA GIPFSAYSAQ SLQHMQYNAQ YSSASTPQYP TAHPLVQAQQ WTW
Greater than 90% as determined by SDS-PAGE.
18088
200 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers