missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse M-CSF / CSF-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) M-CSF / CSF-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8587-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001107001.1 | |
M-CSF / CSF-1 Protein | |
Research Use Only | |
CSF1 | |
Recombinant Protein | |
E. coli | |
KEVSEHCSHM IGNGHLKVLQ QLIDSQMETS CQIAFEFVDQ EQLDDPVCYL KKAFFLVQDI IDETMRFKDN TPNANATERL QELSNNLNSC FTKDYEEQNK ACVRTFHETP LQLLEKIKNF FNETKNLLEK DWNIFTKNCN NSFAKCSSRD VVTKPDCNCL YPKATPSSDP ASASPHQPPA PSMAPLAGLA WDDSQRTEGS SLLPSELPLR IEDPGSAKQR PPRSTCQTLE | |
Greater than 90% as determined by SDS-PAGE. |
12977 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |