missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse M-CSF / CSF-1 Protein

A DNA sequence encoding the Mus musculus (Mouse) M-CSF / CSF-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8587-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015431

  • 3405.00€ / 1mg
Estimated Shipment: 27-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001107001.1
M-CSF / CSF-1 Protein
Research Use Only
CSF1
Recombinant Protein
E. coli
KEVSEHCSHM IGNGHLKVLQ QLIDSQMETS CQIAFEFVDQ EQLDDPVCYL KKAFFLVQDI IDETMRFKDN TPNANATERL QELSNNLNSC FTKDYEEQNK ACVRTFHETP LQLLEKIKNF FNETKNLLEK DWNIFTKNCN NSFAKCSSRD VVTKPDCNCL YPKATPSSDP ASASPHQPPA PSMAPLAGLA WDDSQRTEGS SLLPSELPLR IEDPGSAKQR PPRSTCQTLE
Greater than 90% as determined by SDS-PAGE.
12977
1 mg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers