missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Integrin alpha-L Protein

A DNA sequence encoding the Mus musculus (Mouse) Integrin alpha-L, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6240-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061850

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

Integrin alpha-L Protein
Research Use Only
Itgal
Recombinant Protein
E. coli
DLVFLFDGSQ SLDRKDFEKI LEFMKDVMRK LSNTSYQFAA VQFSTDCRTE FTFLDYVKQN KNPDVLLGSV QPMFLLTNTF RAINYVVAHV FKEESGARPD ATKVLVIITD GEASDKGNIS AAHDITRYII GIGKHFVSVQ KQKTLHIFAS EPVEEFVKIL DTFEKLKDLF TDL
Greater than 90% as determined by SDS-PAGE.
50 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers