missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Integrin alpha-L Protein
A DNA sequence encoding the Mus musculus (Mouse) Integrin alpha-L, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6240-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
Integrin alpha-L Protein | |
Research Use Only | |
Itgal | |
Recombinant Protein | |
E. coli | |
DLVFLFDGSQ SLDRKDFEKI LEFMKDVMRK LSNTSYQFAA VQFSTDCRTE FTFLDYVKQN KNPDVLLGSV QPMFLLTNTF RAINYVVAHV FKEESGARPD ATKVLVIITD GEASDKGNIS AAHDITRYII GIGKHFVSVQ KQKTLHIFAS EPVEEFVKIL DTFEKLKDLF TDL | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |