missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse I-TAC / CXCL11 Protein

A DNA sequence encoding the Mus musculus (Mouse) I-TAC / CXCL11, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8656-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035801

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_062367.1
I-TAC / CXCL11 Protein
Research Use Only
CXCL11
Recombinant Protein
E. coli
FLMFKQGRCL CIGPGMKAVK MAEIEKASVI YPSNGCDKVE VIVTMKAHKR QRCLDPRSKQ ARLIMQAIEK KNFLRRQ
Greater than 90% as determined by SDS-PAGE.
56066
10 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers