missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Hyaluronan synthase 2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Hyaluronan synthase 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6144-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032242.3 | |
Hyaluronan synthase 2 Protein | |
Research Use Only | |
Has2 | |
Recombinant Protein | |
E. coli | |
EHRKMKKSLE TPIKLNKTVA LCIAAYQEDP DYLRKCLQSV KRLTYPGIKV VMVIDGNSDD DLYMMDIFSE VMGRDKSATY IWKNNFHEKG PGETEESHKE SSQHVTQLVL SNKSICIMQK WGGKREVMYT AFRALGRSVD YVQVCDSDTM LDPASSVEMV KVLEEDPMVG GVGGDVQILN KYDSWISFLS SVRYWMAFNI ERACQSYFGC VQCISGPLGM YRNSLLHEFV EDWYNQEFMG NQCSFGDDRH LTNRVLSLGY ATKYTARSKC LTETPIEYLR WLNQQTRWSK SYFREWLYNA MWFHKHHL | |
Greater than 90% as determined by SDS-PAGE. |
15117 | |
200 μg | |
Not determined. | |
Cytoplasmic Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |