missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Hmgcs1, cytoplasmic Protein

A DNA sequence encoding the Mus musculus (Mouse) Hmgcs1, cytoplasmic, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7817-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16060981

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001278368.1
Hmgcs1, cytoplasmic Protein
Research Use Only
Hmgcs1
Recombinant Protein
Yeast
MPGSLPLNAE ACWPKDVGIV ALEIYFPSQY VDQAELEKYD GVDAGKYTIG LGQARMGFCT DREDINSLCL TVVQKLMERH SLSYDCIGRL EVGTETIIDK SKSVKSNLMQ LFEESGNTDI EGIDTTNACY GGTAAVFNAV NWVESSSWDG RYALVVAGDI AIYATGNARP TGGVGAVALL IGPNAPLIFD RGLRGTHMQH AYDFYKPDML SEYPVVDGKL SIQCYLSALD RCYSVYRKKI RAQWQKEGKD KDFTLNDFGF MIFHSPYCKL VQKSLARMFL NDFLNDQNRD KNSIYSGLEA FGDVKLEDTY FDRDVEKAFM KASSELFNQK TKASLLVSNQ NGNMYTSSVY GSLASVLAQY SPQQLAGKRV GVFSYGSGLA ATLYSLKVTQ DATPGSALDK ITASLCDLKS RLDSRTCVAP DVFAENMKLR EDTHHLANYI PQCSIDSLFE GTWYLVRVDE KHRRTYARRP FTNDHSLDEG MGLVHSNTAT EHIPSPAKKV PRLPATSAES ESAVISNGEH
Greater than 90% as determined by SDS-PAGE.
208715
200 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers