missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Glypican-4 Protein

A DNA sequence encoding the Mus musculus (Mouse) Glypican-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6110-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030870

  • 770.00€ / 100µg
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_032176.2
Glypican-4 Protein
Research Use Only
Gpc4
Recombinant Protein
Yeast
AGGAHAEAKP YLLAALCILF LAVQGEWR
Greater than 90% as determined by SDS-PAGE.
14735
100 μg
Not determined.
Partial (see sequence information for more details).
Mouse
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers