missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse E3 ubiquitin-protein ligase Mdm2 Protein
A DNA sequence encoding the Mus musculus (Mouse) E3 ubiquitin-protein ligase Mdm2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6353-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001275515.1 | |
E3 ubiquitin-protein ligase Mdm2 Protein | |
Research Use Only | |
Mdm2 | |
Recombinant Protein | |
Yeast | |
MCNTNMSVST EGAASTSQIP ASEQETLVRP KPLLLKLLKS VGAQNDTYTM KEIIFYIGQY IMTKRLYDEK QQHIVYCSND LLGDVFGVPS FSVKEHRKIY AMIYRNLVAV SQQDSGTSLS ESRRQPEGGS DLKDPLQAPP EEKPSSSDLI SRLSTSSRRR SISETEENTD ELPGERHRKR RRSLSFDPSL GLCELREMCS GGSSSSSSSS SESTETPSHQ DLDDGVSEHS GDCLDQDSVS DQFSVEFEVE SLDSEDYSLS DEGHELSDED DEVYRVTVYQ TGESDTDSFE GDPEISLADY WKCTSCNEMN PPLPSHCKRC WTLRENWLPD DKGKDKVEIS EKAKLENSAQ AEEGLDVPDG KKLTENDAKE PCAEEDSEEK AEQTPLSQES DDYSQPSTSS SIVYSSQESV KELKEETQDK DESVESSFSL NAIEPCVICQ GRPKNGCIVH GKTGHLMSCF TCAKKLKKRN KPCPVCRQPI QMIVLTYFN | |
Greater than 90% as determined by SDS-PAGE. |
17246 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |