missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Cytochrome P450 3A13 Protein
A DNA sequence encoding the Mus musculus (Mouse) Cytochrome P450 3A13, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7720-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_031845.1 | |
Cytochrome P450 3A13 Protein | |
Research Use Only | |
Cyp3a13 | |
Recombinant Protein | |
E. coli | |
MDLIPNFSME TWMLLATSLV LLYLYGTHSH GIFKKLGIPG PKPLPFLGTI LAYQKGFWEC DIQCHKKYGK MWGLYDGRQP VLAITDPDII KTVLVKECYS TFTNRRRFGP VGILKKAISI SENEEWKRIR ALLSPTFTSG RLKEMFPIIN QFTDVLVRNM RQGLGEGKPT SMKDIFGAYS MDVITATSFG VNIDSLNNPQ DPFVEKIKKL LKFDIFDPLF LSVTLFPFLT PVFDALNVSL FPRDVISFFT TSVERMKENR MKEKEKQRVD FLQLMINSQN YKTKESHKAL SDVEIVAQSV IFIFAGYETT SSALSFALYL LAIHPDVQKK LQDEIDAALP NKAPATYDTL LQMEYLDMVV NETLRLYPIA GRLERVCKTD VEINGLFIPK GTVVMIPTFA LHKDPKYWPE PEEFRPERFS KKNQDSINPY MYLPFGSGPR NCIGMRFALI NMKVALVRVL QNFTVQPCKE TEIPLKLSKQ GLLQPENPLL LKVVSRDETV SDE | |
Greater than 90% as determined by SDS-PAGE. |
13113 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |