missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Cystathionine beta-synthase Protein

A DNA sequence encoding the Mus musculus (Mouse) Cystathionine beta-synthase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7956-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16041821

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001258282.1
Cystathionine beta-synthase Protein
Research Use Only
CBS
Recombinant Protein
E. coli
PSGTSQCEDG SAGGFQHLDM HSEKRQLEKG PSGDKDRVWI RPDTPSRCTW QLGRAMADSP HYHTVLTKSP KILPDILRKI GNTPMVRINK ISKNAGLKCE LLAKCEFFNA GGSVKDRISL RMIEDAERAG NLKPGDTIIE PTSGNTGIGL ALAAAVKGYR CIIVMPEKMS MEKVDVLRAL GAEIVRTPTN ARFDSPESHV GVAWRLKNEI PNSHILDQYR NASNPLAHYD DTAEEILQQC DGKLDMLVAS AGTGGTITGI ARKLKEKCPG CKIIGVDPEG SILAEPEELN QTEQTAYEVE GIGYDFIPTV LDRAVVDKWF KSNDEDSFAF ARMLIAQEGL LCGGSSGSAM AVAVKAAREL QEGQRCVVIL PDSVRNYMSK FLSDKWMLQK GFMKEELSVK RPWWWRLRVQ ELSLSAPLTV LPTVTCEDTI AILREKGFDQ APVVNESGAI LGMVTLGNML SSLLAGKVRP SDEVCKVLYK QFKPIHLTDT LGTLSHILEM DHFALVVHEQ IQSRDQAWSG VVGGPTDCSN GMSSKQQMVF GVVTAIDLLN FVAAREQTQT
Greater than 90% as determined by SDS-PAGE.
12411
10 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers