missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD16 / FCGR3 Protein

A DNA sequence encoding the Mus musculus (Mouse) CD16 / FCGR3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7302-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048090

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

CD16 / FCGR3 Protein
Research Use Only
FCGR3
Recombinant Protein
E. coli
ALPKAVVKLD PPWIQVLKED MVTLMCEGTH NPGNSSTQWF HNGRSIRSQV QASYTFKATV NDSGEYRCQM EQTRLSDPVD LGVISDWLLL QTPQRVFLEG ETITLRCHSW RNKLLNRISF FHNEKSVRYH HYKSNFSIPK ANHSHSGDYY CKGSLGSTQH QSKPVTITVQ DPATTSSISL VWYHT
Greater than 90% as determined by SDS-PAGE.
50 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers