Learn More
R&D Systems™ Recombinant Mouse BTNL3 Fc Chimera Protein
Product Code. 18776676
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. 18776676
Brand: R&D Systems™ 10222BT050
Measured by its ability to inhibit IL-2 secretion by mouse T cells in the presence of anti-CD3. The ED50 for this effect is 0.4-4 μg/mL.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q7TST0.3
Conjugate Unconjugated
For Use With (Application) Bioactivity
Gene ID (Entrez) 10917
Quantity 50 μg
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Synonym BTN9.1, BTNLR, BTNLRButyrophilin-like receptor, butyrophilin-like 3, butyrophilin-like protein 3, COLF4100
Target Mouse
Buffer Supplied as a 0.2 μm filtered solution in PBS.