missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse BAG family molecular chaperone regulator 3 Protein
A DNA sequence encoding the Mus musculus (Mouse) BAG family molecular chaperone regulator 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8071-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_038891.4 | |
BAG family molecular chaperone regulator 3 Protein | |
Research Use Only | |
Bag3 | |
Recombinant Protein | |
E. coli | |
SAATQSPMMQ MASGNGASDR DPLPPGWEIK IDPQTGWPFF VDHNSRTTTW NDPRVPPEGP KDTASSANGP SRDGSRLLPI REGHPIYPQL RPGYIPIPVL HEGSENRQPH LFHAYSQPGV QRFRTEAAAA TPQRSQSPLR GGMTEAAQTD KQCGQMPATA TTAAAQPPTA HGPERSQSPA ASDCSSSSSS ASLPSSGRSS LGSHQLPRGY IPIPVIHEQN ITRPAAQPSF HQAQKTHYPA QQGEYQPQQP VYHKIQGDDW EPRPLRAASP FRSPVRGASS REGSPARSGT PVHCPSPIRV HTVVDRPQPM THREPPPVTQ PENKPESKPG PAGPDLPPGH IPIQVIRREA DSKPVSQKSP PPAEKVEVKV SSAPIPCPSP SPAPSAVPSP PKNVAAEQKA APSPAPAEPA APKSGEAETP PKHPGVLKVE AILEKVQGLE QAVDSFEGKK TDKKYLMIEE YLTKELLALD SVDPEGRADV RQARRDGVRK VQTILEKLEQ KAIDVPGQVQ VYELQPSNLE AEQPLQEIMG AVVADKDKKG PENKDPQTES QQLEAKAATP PNPSNPADSA GNLVAP | |
Greater than 90% as determined by SDS-PAGE. |
29810 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |