missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza A H7N7 (Equine / Prague / 1 / 1956) NucleoProtein

A DNA sequence encoding the Influenza A virus (strain A/Equine/Prague/1/1956 H7N7) Influenza A H7N7 (strain A/Equine/Prague/1/1956) Nucleoprotein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7108-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16027410

  • 1765.00€ / 200µg
Estimated Shipment: 26-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Influenza A H7N7 (Equine / Prague / 1 / 1956) NucleoProtein
Research Use Only
NP
Recombinant Protein
E. coli
MASQGTKRPY EQMETGGERQ NATEIRASVG KMVGGIGRFY IQMCTELKLN DYEGRLIQNS ITIEKMVLSA FDERRNKYLE EHPNTGKDPK KTGGPIYRKR EGKWIRELIL YDKEEIRRIW RQANNGEDAT AGLTHLMIWH SNLNDATYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGIGTMV MELIRMIKRG INDRNFWRGE NGRKTRIAYE RMCNILKGKF QTAAQRAMMD QVRESRNPGN AEIEDLIFLA RSALILRGSV AHKSCLPACV YGLIVASGYD FEREGYSLVG VDPFKLLQNS QIFSLIRPNE NPAHKSQLVW MACHSAAFED LRVSSFIRGT KVIPRGQLST RGIQIASNEN METIDSNTLE LRSRYWAIRT KSGGNTSQQK ASAGQISVQP TFSVQRNLPF ERTTIMAAFT GNNEGRTSDM RTEIIRMMEN AKPDDVSFQG RGVFELSDEK ATNPIVPSFD MSKEGSYFFG DNAEEFDN
Greater than 80% as determined by SDS-PAGE.
200 μg
Not determined.
Full Length
Influenza
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers