missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H1N1 (Hickox / 1940) Hemagglutinin Protein
A DNA sequence encoding the Influenza A virus (strain A/Hickox/1940 H1N1) Influenza A H1N1 (strain A/Hickox/1940) Hemagglutinin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7521-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
Influenza A H1N1 (Hickox / 1940) Hemagglutinin Protein | |
Research Use Only | |
HA | |
Recombinant Protein | |
E. coli | |
DTICIGYHAN NSTDTVDTVL EKNVTVTHSV NLLEDSHNGK LCRLKGIAPL QLGKCNIAGW ILGNPECESL LSKRSWSYIA ETPNSENGTC YPGDFADYEE LREQLSSVSS FERFEIFPKE RSWPNHNINI GVTAACSHAG KSSFYKNLLW LTEKDGSYPN LNKSYVNKKE KEVLVLWGVH HPSNIENQKT LYRKENAYVS VVSSNYNRRF TPEIAERPKV RGQAGRMNYY WTLLEPGDTI IFEANGNLIA PWYAFALSRG LGSGIITSNA SMDECDTKCQ TPQGAINSSL PFQNIHPFTI GECPKYVRST KLRMVTGLRN IPSIQS | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |