missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger protein 346 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 346, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6907-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001295142.1 | |
Zinc finger protein 346 Protein | |
Research Use Only | |
Znf346 | |
Recombinant Protein | |
E. coli | |
MEYPAPATVQ AADGGAAGPY SSSELLEGQE PDGVRFDRER ARRLWEAVSG AQPVGREEVE HMIQKNQCLF TNTQCKVCCA LLISESQKLA HYQSKKHANK VKRYLAIHGM ETLKGETKKL DSDQKSSRSK DKNQCCPICN MTFSSPVVAQ SHYLGKTHAK NLKLKQQSTK VEALHQNREM IDPDKFCSLC HATFNDPVMA QQHYVGKKHR KQETKLKLMA RYGRLADPAV TDFPAGKGYP CKTCKIVLNS IEQYQAHVSG FKHKNQSPKT VASSLGQIPM QRQPIQKDST TLED | |
Greater than 80% as determined by SDS-PAGE. |
23567 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |