missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Zinc finger protein 160 Protein

A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 160, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6902-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036650

  • 770.00€ / 100µg
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001096073.1
Zinc finger protein 160 Protein
Research Use Only
ZNF160
Recombinant Protein
E. coli
MALTQVRLTF RDVAIEFSQE EWKCLDPAQR ILYRDVMLEN YWNLVSLGLC HFDMNIISML EEGKEPWTVK SCVKIARKPR TPECVKGVVT DLLRRWKHWL LLLGICCPKP HGRVSSRLRL SRSLGHFFHS AFATFMGVCD KRVGSIF
Greater than 90% as determined by SDS-PAGE.
90338
100 μg
Not determined.
Full Length of Isoform 2
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers