missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human YEATS domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) YEATS domain-containing protein 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6895-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001287879.1 | |
YEATS domain-containing protein 1 | |
Research Use Only | |
YEATS4 | |
Recombinant Protein | |
E. coli | |
MFKRMAEFGP DSGGRVKGVT IVKPIVYGNV ARYFGKKREE DGHTHQWTVY VKPYRNEDMS AYVKKIQFKL HESYGNPLRV VTKPPYEITE TGWGEFEIII KIFFIDPNER PVTLYHLLKL FQSDTNAMLG KKTVVSEFYD EMIFQDPTAM MQQLLTTSRQ LTLGAYKHET EFAELEVKTR EKLEAAKKKT SFEIAELKER LKASRETINC LKNEIRKLEE DDQAKDI | |
Greater than 80% as determined by SDS-PAGE. |
8089 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |