Learn More
enQuireBio™ Recombinant Human VILIP-1 / VSNL1 Protein
A DNA sequence encoding the Homo sapiens (Human) VILIP-1 / VSNL1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6883-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003376.2 | |
VILIP-1 / VSNL1 Protein | |
Research Use Only | |
VSNL1 | |
Recombinant Protein | |
E. coli | |
GKQNSKLAPE VMEDLVKSTE FNEHELKQWY KGFLKDCPSG RLNLEEFQQL YVKFFPYGDA SKFAQHAFRT FDKNGDGTID FREFICALSI TSRGSFEQKL NWAFNMYDLD GDGKITRVEM LEIIEAIYKM VGTVIMMKMN EDGLTPEQRV DKIFSKMDKN KDDQITLDEF KEAAKSDPSI VLLLQCDIQK | |
Greater than 80% as determined by SDS-PAGE. |
7447 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |