missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human VILIP-1 / VSNL1 Protein

A DNA sequence encoding the Homo sapiens (Human) VILIP-1 / VSNL1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6883-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036540

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_003376.2
VILIP-1 / VSNL1 Protein
Research Use Only
VSNL1
Recombinant Protein
E. coli
GKQNSKLAPE VMEDLVKSTE FNEHELKQWY KGFLKDCPSG RLNLEEFQQL YVKFFPYGDA SKFAQHAFRT FDKNGDGTID FREFICALSI TSRGSFEQKL NWAFNMYDLD GDGKITRVEM LEIIEAIYKM VGTVIMMKMN EDGLTPEQRV DKIFSKMDKN KDDQITLDEF KEAAKSDPSI VLLLQCDIQK
Greater than 80% as determined by SDS-PAGE.
7447
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers