missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Vesicle-associated membrane protein 1

A DNA sequence encoding the Homo sapiens (Human) Vesicle-associated membrane protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6874-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046460

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_055047.2
Vesicle-associated membrane protein 1
Research Use Only
VAMP2
Recombinant Protein
E. coli
MSATAATAPP AAPAGEGGPP APPPNLTSNR RLQQTQAQVD EVVDIMRVNV DKVLERDQKL SELDDRADAL QAGASQFETS AAKLKRKYWW KNLKMMIILG VICAIILIII IVYFST
Greater than 90% as determined by SDS-PAGE.
6844
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers