missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UPF0568 protein C14orf166 Protein
A DNA sequence encoding the Homo sapiens (Human) UPF0568 protein C14orf166, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8275-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057123.1 | |
UPF0568 protein C14orf166 Protein | |
Research Use Only | |
C14orf166 | |
Recombinant Protein | |
E. coli | |
MFRRKLTALD YHNPAGFNCK DETEFRNFIV WLEDQKIRHY KIEDRGNLRN IHSSDWPKFF EKYLRDVNCP FKIQDRQEAI DWLLGLAVRL EYGDNAEKYK DLVPDNSKTA DNATKNAEPL INLDVNNPDF KAGVMALANL LQIQRHDDYL VMLKAIRILV QERLTQDAVA KANQTKEGLP VALDKHILGF DTGDAVLNEA AQILRLLHIE ELRELQTKIN EAIVAVQAII ADPKTDHRLG KVGR | |
Greater than 80% as determined by SDS-PAGE. |
51637 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |