missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UDP-glucuronosyltransferase 1-4 Protein

A DNA sequence encoding the Homo sapiens (Human) UDP-glucuronosyltransferase 1-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6870-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16076430

  • 770.00€ / 100µg
Estimated Shipment: 02-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_009051.1
UDP-glucuronosyltransferase 1-4 Protein
Research Use Only
UGT1A4
Recombinant Protein
E. coli
GKVLVVPTDG SPWLSMREAL RELHARGHQA VVLTPEVNMH IKEEKFFTLT AYAVPWTQKE FDRVTLGYTQ GFFETEHLLK RYSRSMAIMN NVSLALHRCC VELLHNEALI RHLNATSFDV VLTDPVNLCG AVLAKYLSIP AVFFWRYIPC DLDFKGTQCP NPSSYIPKLL TTNSDHMTFL QRVKNMLYPL ALSYICHTFS APYASLASEL FQREVSVVDL VSYASVWLFR GDFVMDYPRP IMPNMVFIGG INCANGKPLS QEFEAYINAS GEHGIVVFSL GSMVSEIPEK KAMAIADALG KIPQTVLWRY TGTRPSNLAN NTILVKWLPQ NDLLGHPMTR AFITHAGSHG VYESICNGVP MVMMPLFGDQ MDNAKRMETK GAGVTLNVLE MTSEDLENAL KAVINDKSYK ENIMRLSSLH KDRPVEPLDL AVFWVEFVMR HKGAPHLRPA AHDLTWYQYH SLDVIGFLLA VVLTVAFITF KCCAYGYRKC LGKKGRVKKA HKSKTH
Greater than 90% as determined by SDS-PAGE.
54657
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers