missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UDP-glucose 4-epimerase Protein

A DNA sequence encoding the Homo sapiens (Human) UDP-glucose 4-epimerase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7531-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16059210

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_000394.2
UDP-glucose 4-epimerase Protein
Research Use Only
GALE
Recombinant Protein
E. coli
MAEKVLVTGG AGYIGSHTVL ELLEAGYLPV VIDNFHNAFR GGGSLPESLR RVQELTGRSV EFEEMDILDQ GALQRLFKKY SFMAVIHFAG LKAVGESVQK PLDYYRVNLT GTIQLLEIMK AHGVKNLVFS SSATVYGNPQ YLPLDEAHPT GGCTNPYGKS KFFIEEMIRD LCQADKTWNA VLLRYFNPTG AHASGCIGED PQGIPNNLMP YVSQVAIGRR EALNVFGNDY DTEDGTGVRD YIHVVDLAKG HIAALRKLKE QCGCRIYNLG TGTGYSVLQM VQAMEKASGK KIPYKVVARR EGDVAACYAN PSLAQEELGW TAALGLDRMC EDLWRWQKQN PSGFGTQA
Greater than 90% as determined by SDS-PAGE.
2582
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers