Learn More
enQuireBio™ Recombinant Human UCP1, mitochondrial brown fat Protein
A DNA sequence encoding the Homo sapiens (Human) UCP1, mitochondrial brown fat, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6868-ec-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16016420
  • 2385.00€ / 1mg
Estimated Shipment: 01-07-2024
Specifications
NP_068605.1
UCP1, mitochondrial brown fat Protein
Research Use Only
UCP1
Recombinant Protein
E. coli
GGLTASDVHP TLGVQLFSAG IAACLADVIT FPLDTAKVRL QVQGECPTSS VIRYKGVLGT ITAVVKTEGR MKLYSGLPAG LQRQISSASL RIGLYDTVQE FLTAGKETAP SLGSKILAGL TTGGVAVFIG QPTEVVKVRL QAQSHLHGIK PRYTGTYNAY RIIATTEGLT GLWKGTTPNL MRSVIINCTE LVTYDLMKEA FVKNNILADD VPCHLVSALI AGFCATAMSS PVDVVKTRFI NSPPGQYKSV PNCAMKVFTN EGPTAFFKGL VPSFLRLGSW NVIMFVCFEQ LKRELSKSRQ TMDCAT
Greater than 90% as determined by SDS-PAGE.
7350
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol