missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UCHL5 Protein

A DNA sequence encoding the Homo sapiens (Human) UCHL5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6866-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006410

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001186190.1
UCHL5 Protein
Research Use Only
UCHL5
Recombinant Protein
E. coli
MTGNAGEWCL MESDPGVFTE LIKGFGCRGA QVEEIWSLEP ENFEKLKPVH GLIFLFKWQP GEEPAGSVVQ DSRLDTIFFA KQVINNACAT QAIVSVLLNC THQDVHLGET LSEFKEFSQS FDAAMKGLAL SNSDVIRQVH NSFARQQMFE FDTKTSAKEE DAFHFVSYVP VNGRLYELDG LREGPIDLGA CNQDDWISAV RPVIEKRIQK YSEGEIRFNL MAIVSDRKMI YEQKIAELQR QLAEEPMDTD QGNSMLSAIQ SEVAKNQMLI EEEVQKLKRY KIENIRRKHN YLPFIMELLK TLAEHQQLIP LVEKFEKHFE KTLLGK
Greater than 90% as determined by SDS-PAGE.
51377
200 μg
Not determined.
Full Length of Isoform 4
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers