missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ubiquitin-like-conjugating enzyme ATG10 Protein

A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-like-conjugating enzyme ATG10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8050-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16032371

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001124500.1
Ubiquitin-like-conjugating enzyme ATG10 Protein
Research Use Only
ATG10
Recombinant Protein
E. coli
MEEDEFIGEK TFQRYCAEFI KHSQQIGDSW EWRPSKDCSD GYMCKIHFQI KNGSVMSHLG ASTHGQTCLP MEEAFELPLD DCEVIETAAA SEVIKYEYHV LYSCSYQVPV LYFRASFLDG RPLTLKDIWE GVHECYKMRL LQGPWDTITQ QEHPILGQPF FVLHPCKTNE FMTPVLKNSQ KINKNVNYIT SWLSIVGPVV GLNLPLSYAK ATSQDERNVP
Greater than 80% as determined by SDS-PAGE.
83734
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers