missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tropomodulin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Tropomodulin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6809-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001159588.1 | |
Tropomodulin-1 Protein | |
Research Use Only | |
TMOD1 | |
Recombinant Protein | |
E. coli | |
MSYRRELEKY RDLDEDEILG ALTEEELRTL ENELDELDPD NALLPAGLRQ KDQTTKAPTG PFKREELLDH LEKQAKEFKD REDLVPYTGE KRGKVWVPKQ KPLDPVLESV TLEPELEEAL ANASDAELCD IAAILGMHTL MSNQQYYQAL SSSSIMNKEG LNSVIKPTQY KPVPDEEPNS TDVEETLERI KNNDPKLEEV NLNNIRNIPI PTLKAYAEAL KENSYVKKFS IVGTRSNDPV AYALAEMLKE NKVLKTLNVE SNFISGAGIL RLVEALPYNT SLVEMKIDNQ SQPLGNKVEM EIVSMLEKNA TLLKFGYHFT QQGPRLRASN AMMNNNDLVR KRRLADLTGP IIPKCRSGV | |
Greater than 80% as determined by SDS-PAGE. |
7111 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |