missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transgelin Protein
A DNA sequence encoding the Homo sapiens (Human) Transgelin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8335-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001001522.1 | |
Transgelin Protein | |
Research Use Only | |
TAGLN | |
Recombinant Protein | |
E. coli | |
ANKGPSYGMS REVQSKIEKK YDEELEERLV EWIIVQCGPD VGRPDRGRLG FQVWLKNGVI LSKLVNSLYP DGSKPVKVPE NPPSMVFKQM EQVAQFLKAA EDYGVIKTDM FQTVDLFEGK DMAAVQRTLM ALGSLAVTKN DGHYRGDPNW FMKKAQEHKR EFTESQLQEG KHVIGLQMGS NRGASQAGMT GYGRPRQIIS | |
Greater than 90% as determined by SDS-PAGE. |
6876 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |