missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcriptional enhancer factor TEF-5 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcriptional enhancer factor TEF-5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8032-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003205.2 | |
Transcriptional enhancer factor TEF-5 Protein | |
Research Use Only | |
TEAD3 | |
Recombinant Protein | |
E. coli | |
MNLDQVSKDK ALQSMASMSS AQIVSASVLQ NKFSPPSPLP QAVFSTSSRF WSSPPLLGQQ PGPSQDIKPF AQPAYPIQPP LPPTLSSYEP LAPLPSAAAS VPVWQDRTIA SSRLRLLEYS AFMEVQRDPD TYSKHLFVHI GQTNPAFSDP PLEAVDVRQI YDKFPEKKGG LKELYEKGPP NAFFLVKFWA DLNSTIQEGP GAFYGVSSQY SSADSMTISV STKVCSFGKQ VVEKVETEYA RLENGRFVYR IHRSPMCEYM INFIHKLKHL PEKYMMNSVL ENFTILQVVT SRDSQETLLV IAFVFEVSTS EHGAQHHVYK LVKD | |
Greater than 80% as determined by SDS-PAGE. |
7005 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |