missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transcriptional enhancer factor TEF-5 Protein

A DNA sequence encoding the Homo sapiens (Human) Transcriptional enhancer factor TEF-5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8032-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16012271

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_003205.2
Transcriptional enhancer factor TEF-5 Protein
Research Use Only
TEAD3
Recombinant Protein
E. coli
MNLDQVSKDK ALQSMASMSS AQIVSASVLQ NKFSPPSPLP QAVFSTSSRF WSSPPLLGQQ PGPSQDIKPF AQPAYPIQPP LPPTLSSYEP LAPLPSAAAS VPVWQDRTIA SSRLRLLEYS AFMEVQRDPD TYSKHLFVHI GQTNPAFSDP PLEAVDVRQI YDKFPEKKGG LKELYEKGPP NAFFLVKFWA DLNSTIQEGP GAFYGVSSQY SSADSMTISV STKVCSFGKQ VVEKVETEYA RLENGRFVYR IHRSPMCEYM INFIHKLKHL PEKYMMNSVL ENFTILQVVT SRDSQETLLV IAFVFEVSTS EHGAQHHVYK LVKD
Greater than 80% as determined by SDS-PAGE.
7005
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers