missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcription intermediary factor 1-alpha Protein
A DNA sequence encoding the Homo sapiens (Human) Transcription intermediary factor 1-alpha, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6842-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003843.3 | |
Transcription intermediary factor 1-alpha Protein | |
Research Use Only | |
TRIM24 | |
Recombinant Protein | |
Yeast | |
KKKTEGLVKL TPIDKRKCER LLLFLYCHEM SLAFQDPVPL TVPDYYKIIK NPMDLSTIKK RLQEDYSMYS KPEDFVADFR LIFQNCAEFN EPDSEVANAG IKLENYFEEL LKNLYPEKRF PK | |
Greater than 90% as determined by SDS-PAGE. |
8805 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |