missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcription elongation factor A protein 1
A DNA sequence encoding the Homo sapiens (Human) Transcription elongation factor A protein 2, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_003186.1 |
---|---|
Gene ID (Entrez) | 6919 |
Name | Transcription elongation factor A protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16009550
|
enQuireBio™
QP7582-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment:
Log in to see stock. |
|||||
16029550
|
enQuireBio™
QP7582-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment:
Log in to see stock. |
|||||
16019550
|
enQuireBio™
QP7582-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16039550
|
enQuireBio™
QP7582-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16049550
|
enQuireBio™
QP7582-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16059550
|
enQuireBio™
QP7582-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifica
NP_003186.1 | |
Transcription elongation factor A protein 1 | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
6919 | |
Research Use Only | |
TCEA2 | |
Recombinant Protein | |
E. coli | |
MMGKEEEIAR IARRLDKMVT KKSAEGAMDL LRELKAMPIT LHLLQSTRVG MSVNALRKQS SDEEVIALAK SLIKSWKKLL DASDAKARER GRGMPLPTSS RDASEAPDPS RKRPELPRAP STPRITTFPP VPVTCDAVRN KCREMLTAAL QTDHDHVAIG ADCERLSAQI EECIFRDVGN TDMKYKNRVR SRISNLKDAK NPDLRRNVLC GAITPQQIAV MTSEEMASDE LKEIRKAMTK EAIREHQMAR TGGTQTDLFT CGKCRKKNCT YTQVQTRSSD EPMTTFVVCN ECGNRWKFC | |
Greater than 90% as determined by SDS-PAGE. |